Thermo Scientific Nalgene Storm Water Sampler, Thermo Scientific
Supplier: Thermo Fisher Scientific
1120-1000 1100-1000
30621-304CS 469.47 CAD
30621-304 73521-039
Thermo Scientific Nalgene Storm Water Sampler, Thermo Scientific
Samplers Water Samplers
The Storm Water Sampler is a convenient and affordable device for collecting quaterly storm water grab samples in compliance with EPA sampling requirements
Floating ball valve automatically closes off the sample port after bottle is full to prevent co-mingling with later run-off and volatile analyte loss. Sampling device is intended for one-time use.
Collects a full liter first flush grab sample within the first minutes of storm water outfall flow.
Certifications: Meets EPA NPDES MSGP (Fed. Reg. FRL-6880-5 10/20/00) and most state general permit grab sampling requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable