BOD Nutrient Buffer Pillows, Hach
Supplier: Hach
1486166 1486398
CA14861-66PK 133.43 CAD
CA14861-66 CA14863-98
BOD Nutrient Buffer Pillows, Hach
Environmental Test Kits
For preparation of BOD5 dilution water. SP based.
Each pillow prepares 3 or 19 liters of dilution water.