SP Bel-Art Utility Trays, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F16300-1015 F16300-1216 F16300-1520 F16300-1818 F16300-2020 F16300-2226
47728-018EA 1182.78 CAD
47728-018 47750-854 47750-856 47750-858 47750-860 47750-862 47750-864
SP Bel-Art Utility Trays, Polypropylene, Bel-Art Products, a part of SP
Trays/Dishes/Pots Laboratory Trays/Dishes
Made from polypropylene, these straight-sided trays offer resistance to temperatures up to 135 °C (274 °F).
  • Polypropylene construction
  • Good temperature resistance
  • Straight-sided design
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable