Hardness (Total) Reagent Set, HR, Digital Titrator, Hach
Supplier: Hach
2448100 2448100
CA11014-494EA 224.55 CAD
CA11014-494 97027-042
Hardness (Total) Reagent Set, HR, Digital Titrator, Hach
Environmental Test Kits
Reagent set for the determination of Total Hardness by EDTA Digital Titrator titration (Hach method 8213). Range: 100-4,000 mg/L as CaCO3. Set contains: Buffer Solution (Hardness 1), ManVer® 2 Hardness Indicator Powder Pillows and 0.800 M EDTA Digital Titrator Cartridge for 40 - 100 tests.
  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economica