Stablcal® Turbidity Standards Calibration Kit, 2100P Portable Turbidimeter, Sealed Vials, Hach
Supplier: Hach
Stablcal®
2659410 2659410 2659405 2659405
CA26594-05EA 1076.09 CAD
97009-528 CA26594-05 97009-550 CA97009-550
Stablcal® Turbidity Standards Calibration Kit, 2100P Portable Turbidimeter, Sealed Vials, Hach
Standards Turbidity Standards
Stablcal® Calibration Standards Kit, for 2100P Portable Turbidimeter.
  • Precise and pre-mixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability
  • Saves time and labor
  • Confidence in results
Available as sealed vial format or bottles
Ordering information: Kit includes <0.1, 20, 100 and 800 NTU standards.