VWR® Cuff Tape
Supplier: VWR International
89495-444 89495-446
89495-444CS 2537.09 CAD
89495-444 89495-446
VWR® Cuff Tape
Tapes
This cuff tape is a conformable polyethylene tape used to ensure an effective seal between the glove and garment in cleanroom gowning.
  • Low-outgassing polyethylene fabrication
  • Perforated for easier tearing by hand
  • Wound on a plastic core
Perforations at 3” intervals allow the user to bind and tear at the optimum length to seal the glove. The low-outgassing polyethylene film with acrylic adhesive removes cleanly leaving no adhesive residue and will not tear gloves.