Starfish MonoBlock, Heidolph Radleys
Supplier: Heidolph NA, LLC
015890070 015892040 015890070CA 015890080CA 015890090CA 015890100CA 015890110CA 015890120CA 015890130CA 015890140CA 015890150CA 015892040CA
CA11007-776EA 1059.18 CAD
CA11007-776 CA11008-228 CA11009-026 CA11009-028 CA11009-030 CA11009-032 CA11009-034 CA11009-036 CA11009-038 CA11009-040 CA11009-042 CA11009-196
Starfish MonoBlock, Heidolph Radleys
Block Heaters Blocks
StarFish MonoBlocks are single blocks with multiple wells of the same size; they are ideal for experiments that use the same size/type vessel
Made of anodized aluminum for strong chemical resistance and excellent heat transfer.