WHEATON® MicroLiter μLplate® Snap Caps and μLmats™, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
07-0020B 07-0020G 07-0020MB 07-0020MG 07-0020MN 07-0020MR 07-0020N 07-0020R 07-0021B 07-0021G 07-0021MB 07-0021MG 07-0021MN 07-0021MR 07-0021N 07-0021R 07-0045B 07-0045G 07-0045MB 07-0045MG 07-0045MN 07-0045MR 07-0045N 07-0045R 07-0075B 07-0075G 07-0075MB 07-0075MG 07-0075MN 07-0075MR 07-0075N 07-0075R 07-0070MB 07-0070MG 07-0070MN 07-0070MR 07-0070B 07-0070G 07-0070N 07-0070R
97044-570PK 113.24 CAD
97044-570 97044-588 97044-606 97044-608 97044-610 97044-612 97044-614 97044-632 97044-650 97044-668 97044-686 97044-688 97044-690 97044-692 97044-694 97044-712 97044-740 97044-760 97044-778 97044-782 97044-784 97044-786 97044-788 97044-806 97044-836 97044-854 97044-872 97044-874 97044-876 97044-878 97044-880 97044-898 89203-910 89203-912 89203-914 89203-916 89231-706 89231-724 89231-742 89231-760
WHEATON® MicroLiter μLplate® Snap Caps and μLmats™, DWK Life Sciences
Seals Microplate Seals
A comprehensive line of 96-well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials.
Polypropylene μLplates™ have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples can be converted to 96-well format, by sealing the limited volume insert on the insideMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable