Hamilton Syringes for CTC Analytics Sampler Handlers, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
CL-G100022-3 CL-G10022S-3 CL-G10026S-AS CL-G1022S-3 CL-G250022-3 CL-G25022-3 CL-G25026-AS CL-G2522S-3 CL-G2526S-AS CL-G50026-AS CL-L1.226P-AS CL-L1026S-2 CL-L1026S-AS CL-L526S-AS HS-1.0235 HS-2.5235 HS-5.0235 XT-G10022-3 XT-G10022S-3 XT-G5022S-3 HS-1.0265 HS-2.5265
97050-504EA 260.28 CAD
97050-504 97050-506 97050-507 97050-508 97050-509 97050-510 97050-511 97050-512 97050-513 97050-514 97050-515 97050-516 97050-517 97050-518 97050-519 97050-561 97050-562 97050-563 97050-611 97050-612 97050-613 97050-614 30618-482 30618-484
Hamilton Syringes for CTC Analytics Sampler Handlers, MicroLiter
Syringes Syringes for Chromatography
The design of the C-line is based on the improvements employed in CTC’s X-Type syringe
Excluded are the inert needle material and the specially treated inside of the glass barrel. As with the X-Type syringe the construction is characterized by a direct glass/needle connection that completely avoids sample contact with glue or cement. A long-life polymer compound of the latest generation is used as plunger tip material. Additionally, a new flange design with adjustable plunger protects the tip from squeezing.