Pressure Reactor, Two-Piece With Bottom Outlet, Ace Glass Incorporated
Supplier: Ace Glass
6425-04 6425-06 6425-12 6425-15 6425-19 6425-201 6425-206 6425-211 6425-214 6425-221 6425-226 6425-23
89054-286EA 1051.94 CAD
89054-286 89054-288 89054-290 89054-292 89054-294 89054-296 89054-298 89054-300 89054-302 89054-304 89054-306 89054-308
Pressure Reactor, Two-Piece With Bottom Outlet, Ace Glass Incorporated
Laboratory Reactor Systems
Two-Piece Pressure Reactor System for use at pressures up to 35 PSI at 100 Degrees C
Has threaded bottom outlet with ball joint bottom oultlet connection.
Ordering information: Complete Systems come Supplied With Valves and Ball Joint Bottom Outlet Glass Body, Schott Flange Head With #15 Center Neck,(2) #15 and (1) #7 Side Necks, Addition Funnel, Condenser, Manifold With Gauge and Rupture Disc, Stirring and Temperature Control Assemblies.