Microcentrifuge Tubes, Graduated, with ClickLok™ MCT Seals, Simport Scientific
Supplier: Simport Scientific
T330-7Y T330-7G T330-6B T330-6G T330-6Y T330-7B T330-7Y T330-72B T330-72G T330-72Y T330-72AB T330-72AG T330-72AY T330-15N T330-15N T330-6B T330-6G T330-6Y T330-72AG T330-72AY T330-72B T330-72G T330-72Y T330-7AM T330-7B T330-8B T330-8G T330-8Y
CA60829-035CS 631.69 CAD
CA60829-035 CA73960-306 CA73960-308 89499-274 89499-276 89499-278 89499-280 89499-282 89499-284 89499-286 89499-288 89499-290 89499-292 89499-294 89499-296 89499-298 89499-300 89499-302 89499-304 89499-306 89499-308 89499-310 89499-322 89499-324 89499-326 89499-328 CA11031-922 CA11031-924 CA11031-926 CA11031-928 CA11031-932 CA11031-934 CA11031-938 CA11031-940 CA11031-946 CA11031-948 CA11031-950 CA11031-952 CA11031-954 CA11031-956 CA11031-960 CA11031-962 CA11031-964 CA11031-966 CA11031-968 CA11031-970 CA11031-972
Microcentrifuge Tubes, Graduated, with ClickLok™ MCT Seals, Simport Scientific
Tubes Microcentrifuge Tubes
These polypropylene microcentrifuge tubes are able to withstand the stress of high speed centrifugation up to 20000×g.
  • With external graduation and frosted writing surfaces
  • Autoclavable to 121 °C
  • Pierceable lid
60872-600 incorporates convenient pick-up tabs for easier handling without actually touching the tube. 89499-322 offers a special locking system which can be bent upward to lock on the cap, ensuring extra protection during critical steps.
The one-piece construction incorporates a snug fitting and has a reliable attached cap, even with prolonged boiling. The bottom is reinforced for added protection against leakage.
Certifications: Certified RNase-, DNase-, Pyrogen- and DNA-free.
Ordering information: Packaged in tamperproof, resealable safety-lock bags.