SP Bel-Art Heavy Duty Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18855-0000 F18856-0000 F18857-0000 F18859-0000
60916-305CS 663.3 CAD
60916-305 60916-349 60916-382 60916-429
SP Bel-Art Heavy Duty Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
These heavy-duty 3 tier racks are stackable and compact, and have hand grips in the end plates for easy transport.
  • Compact design
  • Polypropylene construction
  • Autoclavable
  • Stackable
The base tier has rounded wells with drain holes, and the top tier has lettered and numbered rows. Molded of polypropylene they are steam autoclavable at 121ºC (250ºF). All racks are 21.3 cm (8³/₈") long x 11 cm (4⁵/₁₆") wide.