Solvent Distillation Head, Ace Glass Incorporated
Supplier: Ace Glass
7812-08 7812-10 7812-14
89428-140EA 1323.26 CAD
89428-140 89428-142 89428-144
Solvent Distillation Head, Ace Glass Incorporated
Distillation Components Distillation Heads
This is a borosilicate glass solvent distillation head designed to maintain solvents or vapors in an no-air atmosphere
Top takeoff joint is standard taper 14/20 on all members of this product family.