Tubing Connector with Quick Connect/Disconnect, Ace Glass Incorporated
Supplier: Ace Glass
12517-40 12517-14 12517-08
89083-350EA 48.05 CAD
89083-350 89083-352 89083-354
Tubing Connector with Quick Connect/Disconnect, Ace Glass Incorporated
Fittings and Connectors
This is a two-piece tubing connector, used to connect tubing from a pressure source to the manifold of the ACE pressure reactor
Offers the convenience of a quick connect/disconnect coupling. Body has MPT thread for connecting to an adapter at end of ACE pressure manifold. Coupling insert is connected to tubing from pressure source. Push coupling insert into coupling body for leak-tight connection; press thumb latch to quick disconnect. Fabricated of polypropylene.
Ordering information: Order body and insert separately.