Vacuum Trap With Tube Connections, Bent Top Connection, Ace Glass Incorporated
Supplier: Ace Glass
8751-12 8751-16 8751-04 8751-08 8751-20
89063-898EA 224.66 CAD
89063-898 89063-900 89063-904 89063-906 89063-912
Vacuum Trap With Tube Connections, Bent Top Connection, Ace Glass Incorporated
Vacuum Traps
Used for freezing out undesired vapors in the vacuum system and also for use in conjunction with Dewar flasks
Top is bent in line with side tube. Fabricated from borosilicate glass.