Vacuum Trap With Tube Connections, Ace Glass Incorporated
Supplier: Ace Glass
8750-04 8750-08 8750-20 8750-12 8750-16
89063-894EA 183.96 CAD
89063-894 89063-896 89063-902 89063-908 89063-910
Vacuum Trap With Tube Connections, Ace Glass Incorporated
Vacuum Traps
Used for freezing out undesired vapors in the vacuum system and also for use in conjunction with Dewar flasks
Made from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable