VACUSIP Bench-Top Aspiration System, INTEGRA Biosciences
Supplier: INTEGRA Biosciences
159000 159010
89498-930EA 3944.87 CAD
89498-930 89498-932
VACUSIP Bench-Top Aspiration System, INTEGRA Biosciences
Pumps Vacuum Aspiration Systems
VACUSIP provides an out-of-the-box solution for the wide array of liquid waste disposal tasks carried out at the laboratory bench
  • Reliable low noise pump, enabling it to operate independently from an external vacuum source
  • Rechargeable battery version provides even more flexibility
  • Unique hand operator allows a fine regulation of the aspiration flow and accepts a variety of adapters
The 0.5 L glass bottle is quickly exchanged and all components coming in contact with liquids are autoclavable.
Battery capacity (when fully charged): 5 days when aspirating 1 h per day (model 89498-930 with battery only).
Certifications: CE certified.
Ordering information: Aspiration system with borosilicate bottle, silicone hand operator (incl. 1- and 8-channel steel tips and 1-channel adapter for standard tips/GRIPTIPS), 1.2 m silicone tubing, filter and mains adapter.