Round-Bottom Vacuum Storage Flasks, with Hi-Vac Top Valve and #15 O-Ring Joint Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7816-02 7816-04 7816-06 7816-08
89428-156EA 366.3 CAD
89428-156 89428-158 89428-160 89428-162
Round-Bottom Vacuum Storage Flasks, with Hi-Vac Top Valve and #15 O-Ring Joint Sidearm, Ace Glass Incorporated
Flasks Round Bottom Flasks
This borosilicate glass round bottom flask for vacuum applications
Ideal for storage or reactions. 0-4 mm Hi-Vac valve for precise control of flow. 15 mm O-Ring joint side arm for connecting to schlenk lines. FETFE O-Ring included.