Round-Bottom Two-Neck Flasks, with Sidearm Stopcock or Valve, Ace Glass Incorporated
Supplier: Ace Glass
7799-02 7799-04 7799-08 7799-100 7799-110 7799-112 7799-114
89428-124EA 373.3 CAD
89428-124 89428-126 89428-130 89428-132 89428-134 89428-136 89428-138
Round-Bottom Two-Neck Flasks, with Sidearm Stopcock or Valve, Ace Glass Incorporated
Flasks Round Bottom Flasks
These round-bottom, borosilicate flasks feature two necks and a side port with either a 2 mm glass stopcock or a 0-3 inlet valve
Excellent flask for reactions and NO-AIR or vacuum applications. Side port is available with either a 2 mm glass stopcock or a 0-3 inlet valve with an 8mm end for use with a rubber septa.
Order Now
SPECIFICATIONS
Environmentally Preferable