Pressure Equalizing Funnels, Graduated with PTFE Metering Valve, Ace Glass Incorporated
Supplier: Ace Glass
7320-08 7320-10 7320-12 7320-14 7320-17
89058-776EA 802.41 CAD
89058-776 89058-778 89058-780 89058-782 89058-784
Pressure Equalizing Funnels, Graduated with PTFE Metering Valve, Ace Glass Incorporated
Funnels Separating Funnels
Cylindrical with standard taper joint top and bottom, drip stem, pressure equalizing arm and double scale graduations, calibrated from stopcock
Drip stem does not extend beyond joint, thereby eliminating possible source of breakage. Stopcock available with glass plug, 1:5 PTFE plug or PTFE metering plug. Fabricated from borosilicate glass.
Ordering information: Stoppers are not supplied. For stoppers or replacement stopcock plugs, contact your Avantor representative.