Jacketed One-Piece Pressure Reactor System, Ace Glass Incorporated
Supplier: Ace Glass
6438-247 6438-240 6438-347 6438-353 6438-253 6438-340 6438-350 6438-250 6438-255 6438-355
89054-436EA 16772.35 CAD
89054-436 10767-780 89054-438 10767-288 10767-784 10767-782 89054-440 89054-442 10767-786 89054-434
Jacketed One-Piece Pressure Reactor System, Ace Glass Incorporated
Condensers
Complete, one-piece, jacketed round bottom flask, pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis
Reactor capacities range from 500 mL to 5000 mL and use Ace-Threds to achieve a leak-tight system. One-piece design features a 25 mm center opening and has a higher allowable pressure rating than our two-piece design. Rated 45 PSI at 100 degrees centigrade. Systems are available for either mechanical or magnetic stirring. Jacket is availble with 28/15 O-Ring ball joint or Ace-Thred inlets/outlets. Stand Not Included.
Ordering information: Comes complete with (2) #15 PTFE Plugs, Condenser, Flask, Head, Manifold, Thermowell, Clamp, Funnel, (2) #15 PTFE Bushing, Bearing w/ Ace-Threads, Stir Shaft, Flexible Shaft, PTFE Stirrer Blade, Krytox® High Vacuum Grease, Swivel Coupling, Polypropylene Tubing, Motor Controller, Mounting Rod, PTFE Sealing Tape. Stand not Included.