Jacketed One-Piece Pressure Reactor, Ace Glass Incorporated
Supplier: Ace Glass
6438-14 6438-17 6438-19 6438-24 6438-29 6438-314 6438-317 6438-319 6438-324 6438-329
89054-426EA 3871.98 CAD
89054-426 89054-428 89054-430 89054-432 89054-454 10098-862 10098-864 10098-866 10098-868 10098-870
Jacketed One-Piece Pressure Reactor, Ace Glass Incorporated
Condensers
Rugged heavy wall jacketed one-piece pressure reactor, for use at pressure of up to 45 PSIG @ 100°C
The inlet/outlet on the jacket has easy connect/disconnect size 28/15 O-ring ball joints. Designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Reactor capacities range from 500 mL to 5000 mL and use Ace-Threds to achieve a leak-tight system. One-piece design features a 25 mm center opening and has a higher allowable pressure rating than our two-piece design.