Sparger, Straight with Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
6453-105 6453-04 6453-122 6453-21 6453-107 6453-06 6453-124 6453-23 6453-133 6453-32 6453-146 6453-45 6453-08 6453-126 6453-25
89084-600EA 513.79 CAD
89084-600 89084-602 89084-608 89084-610 89084-616 89084-618 89084-624 89084-626 89084-630 89084-634 89084-640 89084-642 89084-648 89084-650 89084-656 89084-658
Sparger, Straight with Hose Connection, Ace Glass Incorporated
Stirrers Stirring Tools
Straight sparger tube with Ace fritted bottom available in various porosities
Top is either a 1/2" ID hose connection or a #7 Ace-Thred which allows the use of our Ace-Safe connector (89052-936). The 10 mm OD spargers will fit inside #11 Ace-Threds, the 14 mm OD within #15 Ace-Threds.