Jacketed One-Piece Pressure Reactor with Bottom Outlet, Ace Glass Incorporated
Supplier: Ace Glass
6439-15 6439-21 6439-243 6439-248 6439-25 6439-252 6439-254 6439-257 6439-29 6439-34 6439-315 6439-321 6439-325 6439-329 6439-334 6439-343 6439-348 6439-352 6439-354 6439-357
89054-456EA 4412.23 CAD
89054-456 89054-458 89054-460 89054-462 89054-464 89054-466 89054-468 89054-470 89054-472 89054-474 10098-872 10098-874 10098-876 10098-878 10098-880 10767-788 10767-790 10767-792 10767-794 10767-796
Jacketed One-Piece Pressure Reactor with Bottom Outlet, Ace Glass Incorporated
Condensers
Rugged heavy wall jacketed one-piece pressure reactor with bottom outlet valve, for use at pressure of up to 45 PSIG @ 100°C
The inlet/outlet on the jacket has easy connect/disconnect size 28/15 O-ring ball joints. Designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Reactor capacities range from 500ml to 5000ml and use Ace-Threds to achieve a leak-tight system. One-piece design features a 25mm center opening and has a higher allowable pressure rating than our two-piece design.
Order Now
SPECIFICATIONS
Environmentally Preferable