Round-Bottom Vacuum Storage Flasks, with Hi-Vac Top Valve and ⅜" Cajon Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7814-02 7814-04 7814-06 7814-08 7814-10
89428-146EA 331.24 CAD
89428-146 89428-148 89428-150 89428-152 89428-154
Round-Bottom Vacuum Storage Flasks, with Hi-Vac Top Valve and ⅜" Cajon Sidearm, Ace Glass Incorporated
Flasks Round Bottom Flasks
This borosilicate storage tube comes with a 0-4mm Hi-Vac valve top with a 9mm side port designed to accept ⅜" Cajon fittings
The valved top gives the researcher precise flow control.