Single Tube Vacuum Manifold, O-Ring Joint Connections and Easy Action Valves, Ace Glass Incorporated
Supplier: Ace Glass
8745-12 8745-14 8745-31 8745-35 8745-10 8745-33
89084-258EA 1602.55 CAD
89084-258 89084-246 89084-276 89084-264 89084-252 89084-270
Single Tube Vacuum Manifold, O-Ring Joint Connections and Easy Action Valves, Ace Glass Incorporated
Manifolds
The no.15 o-ring joint connections at each end
Three or four ports with Easy-Action 0 - 10 mm threaded stopcocks and standard taper joints or 15 o-ring joint connections off each port. The distance between ports is 95 mm, which will accept 250 ml flasks or smaller. Requires all stainless steel, pinch type screw locking clamp 89060-256 (for 15 o-ring joint).