All-In-One Hardware Kits (use with OPTI-LYNX™), Optimize Technologies
Supplier: Optimize
11-02323-AA 11-02325-AA
77100-348EA 2037.69 CAD
77100-348 77100-352
All-In-One Hardware Kits (use with OPTI-LYNX™), Optimize Technologies
Chromatography Columns
Opti-Lynx inserts can be a filter, guard column, trap cartridge or analytical column.
Since the interacting surfaces are identical, the only variable component is the holder tube. Install a guard column for one application and then switch to a pre-column filter for the next. Use an in-line filter with different porosities and diameters just by changing the insert. Switch out columns in an instant with the Velocity® system or retrofit current columns with the universal port adapter. You can make and break flow paths for multiple detectors, or anywhere else a break in flow is frequently made. Opti-Lynx allows all of this without ever having to change out the basic components.