Round Bottom Flask with Duran Flange Center Neck and Angled Side Port, Ace Glass Incorporated
Supplier: Ace Glass
6530-42 6530-43 6530-44 6530-47 6530-48 6530-49 6530-52 6530-54 6530-56 6530-64 6530-65 6530-66
89055-380EA 7408.67 CAD
89055-380 89055-382 89055-384 89055-388 89055-390 89055-392 89055-396 89055-398 89055-400 89055-404 89055-406 89055-408
Round Bottom Flask with Duran Flange Center Neck and Angled Side Port, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Rugged, spherical reaction flask with Duran style o-ring grooved side port, angled 45° for ease in adding material to flask
In addition to the side port, flask has three standard taper 45/50 reinforced side necks, all vertical. Center is a Duran style o-ring grooved flange supplied with a CAPFE (PTFE encapsulated silicone) o-ring for use with quick release clamp (not included). Supplied with capacity reference gradations. Order cap for side port, clamp for side port, clamp for center flange, and extra o-rings separately.