OPTI-LYNX™ Trap Cartridges
Supplier: Optimize
11-02868-TK 11-02868-TL 11-02868-DQ 11-03787-DQ 11-03787-TA 11-03787-TB 11-03787-TD 11-03787-TE 11-03787-TF 11-03787-TG 11-03787-TH 11-03787-TK 11-03787-TL 11-02870-DQ 11-02870-TK 11-02870-TL 11-02872-DQ 11-02872-TK 11-02872-TL 11-02874-DQ 11-02874-TK 11-02874-TL 11-02868-TD 11-02868-TE 11-02868-TF 11-02868-TG 11-02868-TH 11-02870-TD 11-02870-TE 11-02870-TF 11-02870-TG 11-02870-TH 11-02872-TD 11-02872-TE 11-02872-TF 11-02872-TG 11-02872-TH 11-02874-TD 11-02874-TE 11-02874-TF 11-02874-TG 11-02874-TH
89207-644PK 2120.79 CAD
89207-644 89207-646 89207-648 89207-650 89207-652 89207-654 89207-656 89207-658 89207-660 89207-662 89207-664 89207-666 89207-668 89207-670 89207-672 89207-674 89207-676 89207-678 89207-680 89207-682 89207-684 89207-686 89522-270 89522-272 89522-274 89522-276 89522-278 89522-280 89522-282 89522-284 89522-286 89522-288 89522-290 89522-292 89522-294 89522-296 89522-298 89522-300 89522-302 89522-304 89522-306 89522-308
OPTI-LYNX™ Trap Cartridges
Chromatography Columns
OPTI-LYNX™ trap cartridges combine the convenience of our quick-connect holder with a versatile selection of packed-bed cartridges to offer chromatographers numerous options for on-line or off-line sample clean-up and pre-concentration.
  • Rated to 6000 psi
  • Bed volumes from 4 to 100 µl (perfect for large volume trapping applications)
  • Can be loaded and regenerated
  • Cartridges share the same external dimensions
  • Can be used in on-line and off-line applications
Each cartridge shares the same external dimensions, allowing you to use different diameters andMore Product Information