Funnels, Pressure Equalizing, Graduated with Glass Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7296-50 7296-52 7296-54 7296-56 7296-58
89058-726EA 712.96 CAD
89058-726 89058-728 89058-730 89058-732 89058-734
Funnels, Pressure Equalizing, Graduated with Glass Stopcock, Ace Glass Incorporated
Funnels Dropping Funnels
Cylindrical with standard taper joint top and bottom, drip stem, pressure equalizing arm and double scale graduations, calibrated from stopcock
Drip stem does not extend beyond joint, thereby eliminating possible source of breakage.