Two-Piece Jacketed Pressure Reactor without Drain Valve, Ace Glass Incorporated
Supplier: Ace Glass
6427-09 6427-13 6427-16 6427-18 6427-207 6427-209 6427-212 6427-217 6427-223 6427-228 6427-23 6427-26 6427-264 6427-266 6427-268 6427-270 6427-272 6427-274 6427-309 6427-313 6427-316 6427-318 6427-323 6427-326 6427-428 6427-407 6427-409 6427-412 6427-417 6427-423
89054-310EA 2790.7 CAD
89054-310 89054-312 89054-314 89054-316 89054-318 89054-320 89054-322 89054-324 89054-326 89054-328 89054-330 89054-332 89054-334 89054-336 89054-338 89054-340 89054-342 89054-344 10098-836 10098-838 10098-840 10098-842 10098-844 10098-846 10767-274 10767-770 10767-772 10767-774 10767-776 10767-778
Two-Piece Jacketed Pressure Reactor without Drain Valve, Ace Glass Incorporated
Laboratory Reactor Systems
Complete, two-piece, jacketed flask pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis, and catalysis
Rated 36 psi at 100 °C. Maximum jacket pressure is 8 psig.
Reactor capacities range from 500 mL to 5000 mL, and use Ace-Threds to achieve a leak-tight system. Two-piece (Duran® flanged flask and head) design allows the use of large agitators and facilitates product removal and clean-out. Systems can be selected with either a shaft and agitator style mixing, or stir bar mixing with the use of a super magnet suspended below the reactor body. Reactor bodies are available with 28/15 ball joint or Ace-Thred inlet and outlets. Does not include stand.
Ordering information: System is supplied complete with (2) #15 PTFE plugs, condenser, flask, head, manifold, thermowell, clamp, funnel, (2) #15 PTFE bushings, bearing with Ace-Threds, stir shaft, flexible shaft, PTFE stirrer blade, Krytox high vacuum grease, swivel coupling, polypropylene tubing, motor controller and selected mixing type, mounting rod, and PTFE sealing tape. Stir bar style systems come with raised ring octaganol stirrer magnet.