Polyvinyl alcohol, M.W. ∼200000 for synthesis, partially hydrolysed, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

M-Clarity™ quality level = MQ200

8.43871.0100 8.43871.1000 8.43871.0100 8.43871.1000
EM8.43871.0100CS 1079.14 CAD
EM8.43871.0100 EM8.43871.1000 CA8.43871.0100 CA8.43871.1000
Polyvinyl alcohol, M.W. ∼200000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: >200 °C
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00081922
CAS Number: 9002-89-5
EINECS: 207-767-2

Order Now

Specification Test Results

Storage temperature 15...25 °C
Degree of hydrolysis ≥85%
dyn.Viscosity 34,0 - 46,0 mPa∙s
Loss on drying (110 °C, 2 h) ≤5,0%
Ash ≤1,0%
Ester value 130 - 150


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR