Polyvinyl alcohol, M.W. ∼ 160000 for synthesis, partially hydrolysed, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

M-Clarity™ quality level = MQ200

8.43870.1000 8.43870.0100 8.43870.1000
EM8.43870.0100CS 1061.76 CAD
EM8.43870.0100 EM8.43870.1000 CA8.43870.0100 CA8.43870.1000
Polyvinyl alcohol, M.W. ∼ 160000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 200 °C
Bulk Density 400 - 600 kg/m3
Density 1.3 g/cm3 (20 °C)
pH 4.5 - 7.0 (40 g/l, H2O, 23 °C)
Ignition Point 230 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR