Polyvinyl alcohol, M.W. ∼ 60000 for synthesis, fully hydrolysed, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43866.0100 8.43866.1000 8.43866.0100 8.43866.1000
EM8.43866.0100CS 1104.42 CAD
EM8.43866.0100 EM8.43866.1000 CA8.43866.0100 CA8.43866.1000
Polyvinyl alcohol, M.W. ∼ 60000 for synthesis, fully hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Melting Point 160 - 240 °C
Bulk Density 400 - 600 kg/m3
Density 0.4 - 0.6 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR