tert-Butyl ethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41873.0002 8.41873.0005 8.41873.0025 8.41873.0002 8.41873.0005 8.41873.0025
EM8.41873.0002CS 3818.86 CAD
EM8.41873.0002 EM8.41873.0005 EM8.41873.0025 CA8.41873.0002 CA8.41873.0005 CA8.41873.0025
tert-Butyl ethyl ether for synthesis, Sigma-Aldrich®
tert-Butyl ethyl ether
Formula: C₆H₁₄O
Boiling Pt: 71 °C (1013 hPa)
Melting Pt: –97 °C
Density: 0.752 g/cm³ (20 °C)
MDL Number: MFCD00009225
CAS Number: 637-92-3
EINECS: 211-309-7
UN: 1179
ADR: 3,II
Merck Index: 13,03809

Order Now

Specification Test Results

Solubility 12 g/l (25 °C)
Melting Point -97 °C
Molar Mass 102.18 g/mol
Boiling Point 71 °C (1013 hPa)
Vapor Pressure 173 hPa (25 °C)
Flash Point -19 °C
Explosion Limit 1.23 - 7.7 %(V)
Density 0.74 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR