Calcium hypochlorite for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41799.0100 8.41799.0500 8.41799.0100 8.41799.0500
EM8.41799.0100CS 1575.26 CAD
EM8.41799.0100 EM8.41799.0500 CA8.41799.0100 CA8.41799.0500
Calcium hypochlorite for synthesis, Sigma-Aldrich®
Calcium hypochlorite
Formula: CaCl₂O₂
Melting Pt: ∼100 °C
Density: 2.35 g/cm³ (20 °C)
MDL Number: MFCD00010900
CAS Number: 7778-54-3
EINECS: 231-908-7
UN: 1748
ADR: 5.1,II
Merck Index: 13,02111

Order Now

Specification Test Results

Assay (iodometric) 67.0 - 75.0 %
Identity passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR