Diethyl pyrocarbonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

M-Clarity™ quality level = MQ200

8.41751.0005 8.41751.0025 8.41751.0005 8.41751.0025
EM8.41751.0005CS 4324.46 CAD
EM8.41751.0005 EM8.41751.0025 CA8.41751.0005 CA8.41751.0025
Diethyl pyrocarbonate for synthesis, Sigma-Aldrich®
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93 °C (1013 hPa)
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now

Specification Test Results

Molar Mass 162.14 g/mol
Boiling Point 93 - 94 °C (24 hPa)
Flash Point 69 °C
Density 1.12 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR