Sodium 1-decanesulfonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.41036.0010
8.41036.0050
8.41036.0010
8.41036.0050
8.41036.0002
EM8.41036.0010CS
10404.3
CAD
EM8.41036.0010
EM8.41036.0050
CA8.41036.0010
CA8.41036.0050
CA8.41036.0002
Sodium 1-decanesulfonate for synthesis, Sigma-Aldrich®
Sodium 1-decanesulphonate
Formula:
C₁₀H₂₁NaO₃S Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007526 CAS Number: 13419-61-9 EINECS: 236-525-9 |
Specification Test Results
Assay (acidimetric after ion exchange) | ≥ 98.0 % |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...