4-Vinylbenzoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41019.0001 8.41019.0001
EM8.41019.0001CS 18728.12 CAD
EM8.41019.0001 CA8.41019.0001
4-Vinylbenzoic acid for synthesis, Sigma-Aldrich®
4-Vinylbenzoic acid
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00002569
CAS Number: 1075-49-6

Order Now

Specification Test Results

Molar Mass 148.16 g/mol
Density 1.30 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR