Span® 65, Span® for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sorbitan tristearate

M-Clarity™ quality level = MQ200

8.40122.0100 8.40122.0100 8.40122.0500
EM8.40122.0100CS 4866.4 CAD
EM8.40122.0100 CA8.40122.0100 CA8.40122.0500
Span® 65, Span® for synthesis, Sigma-Aldrich®
Span® 65
Formula: C₆₀H₁₁₄O₈
MW: 963.56 g/mol
Density: 0.98 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 26658-19-5
EINECS: 247-891-4

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 53 °C
Molar Mass 963.55 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure  - 1.4 hPa (20 °C)
Flash Point 149 °C
Density 0.98 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR