1-Cetylpyridinium chloride monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate

M-Clarity™ quality level = MQ200

8.40008.0025 8.40008.0100 8.40008.0025 8.40008.0100
EM8.40008.0025CS 11376 CAD
EM8.40008.0025 EM8.40008.0100 CA8.40008.0025 CA8.40008.0100
1-Cetylpyridinium chloride monohydrate for synthesis, Sigma-Aldrich®
1-Cetylpyridinium chloride monohydrate
Formula: C₂₁H₃₈ClN·1H₂O
MW: 358.01 g/mol
Melting Pt: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS Number: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1 I
Merck Index: 13,02041

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point 217 °C
Molar Mass 358.01 g/mol
Bulk Density 370 kg/m3
pH 5.0 - 5.4 (10 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR