Acetanilide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22344.0100 8.22344.0500 8.22344.0100 8.22344.0500
EM8.22344.0100CS 916.4 CAD
EM8.22344.0100 EM8.22344.0500 CA8.22344.0100 CA8.22344.0500
Acetanilide for synthesis, Sigma-Aldrich®
Acetanilide
Formula: C₈H₉NO
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now

Specification Test Results

Solubility 5 g/l (20 °C)
Melting Point 115 °C
Molar Mass 135.16 g/mol
Bulk Density 370 kg/m3
Boiling Point 304 °C (1013 hPa)
Vapor Pressure 0.002 hPa (20 °C)
Flash Point 173 °C DIN 51758
Density 1.22 g/cm3 (20 °C)
pH 5 - 7 (10 g/l, H2O, 25 °C)
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR