Sodium azide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.22335.0250 8.22335.1000 8.22335.0100 8.22335.0250 8.22335.1000
EM8.22335.0250CS 2219.9 CAD
EM8.22335.0250 EM8.22335.1000 CA8.22335.0100 CA8.22335.0250 CA8.22335.1000
Sodium azide for synthesis, Sigma-Aldrich®
Sodium azide
Formula: NaN₃
Melting Pt: 275 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 13,08653

Order Now

Specification Test Results

Solubility 420 g/l (17 °C)
Melting Point 275 °C (decomposition)
Molar Mass 65.01 g/mol
Boiling Point 300 °C (rigorous decomposition)
Density 1.85 g/cm3 (20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR