(±)-Tetrahydrofurfuryl alcohol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA

M-Clarity™ quality level = MQ200

8.22307.0100 8.22307.1000 8.22307.2500 8.22307.0100 8.22307.1000 8.22307.2500
EM8.22307.0100CS 951.16 CAD
EM8.22307.0100 EM8.22307.1000 EM8.22307.2500 CA8.22307.0100 CA8.22307.1000 CA8.22307.2500
(±)-Tetrahydrofurfuryl alcohol for synthesis, Sigma-Aldrich®
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: < –80 °C
Density: 1.0543 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 102.13 g/mol
Boiling Point 178 °C (1013 hPa)
Vapor Pressure 1 hPa (25 °C)
Flash Point 75 °C
Explosion Limit 1.5 - 9.7 %(V)
Density 1.05 g/cm3 (20 °C)
Ignition Point 282 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR