Resorcinol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,3-Dihydroxybenzene, 1,3-Benzenediol

M-Clarity™ quality level = MQ200

8.22303.0250 8.22303.1000 8.22303.0250 8.22303.1000
EM8.22303.0250CS 4461.92 CAD
EM8.22303.0250 EM8.22303.1000 CA8.22303.0250 CA8.22303.1000
Resorcinol for synthesis, Sigma-Aldrich®
Resorcinol
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 281
Melting Pt: 109…111 °C
Density: 1.272 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002269
CAS Number: 108-46-3
EINECS: 203-585-2
UN: 2876
ADR: 6.1 III
Merck Index: 12,08323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point 109 - 111 °C
Molar Mass 110.11 g/mol
Bulk Density 600 - 700 kg/m3
Boiling Point 281 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 171 °C
Explosion Limit 1.4 %(V)
Density 1.28 g/cm3 (20 °C)
pH 4 - 6 (100 g/l, H2O, 20 °C)
Ignition Point 605 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR