1-Naphthol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene

M-Clarity™ quality level = MQ200

8.22289.0250 8.22289.1000 8.22289.0250 8.22289.1000
EM8.22289.0250CS 638.32 CAD
EM8.22289.0250 EM8.22289.1000 CA8.22289.0250 CA8.22289.1000
1-Naphthol for synthesis, Sigma-Aldrich®
1-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 282
Melting Pt: 94
Density: 1.224 g/cm³ (20 °C)
Flash Pt: 125
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,06409

Order Now

Specification Test Results

Solubility 0.1 g/l (20 °C)
Melting Point 94 - 96 °C
Molar Mass 144.17 g/mol
Bulk Density 450 kg/m3
Boiling Point 288 °C (1013 hPa)
Vapor Pressure 1.3 hPa (94 °C)
Flash Point 125 °C
Density 1.28 g/cm3 (20 °C)
Ignition Point 510 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR