Sodium sulfate, anhydrous for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.22286.1000
8.22286.5000
8.22286.1000
8.22286.5000
EM8.22286.1000CS
540.36
CAD
EM8.22286.1000
EM8.22286.5000
CA8.22286.1000
CA8.22286.5000
Sodium sulfate, anhydrous for synthesis, Sigma-Aldrich®
Sodium sulphate
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 888 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 12,08829 |
|
Specification Test Results
Solubility | 44.55 g/l (20 °C) |
Melting Point | 888 °C |
Molar Mass | 142.04 g/mol |
Bulk Density | 1400 - 1600 kg/m3 |
Density | 2.70 g/cm3 (20 °C) |
pH | 4 - 7 (200 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...