Sodium nitrite, crystals for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.22285.1000
8.22285.5000
8.22285.1000
8.22285.5000
EM8.22285.0100CS
4427.98
CAD
EM8.22285.0100
EM8.22285.1000
EM8.22285.5000
CA8.22285.0100
CA8.22285.1000
CA8.22285.5000
Sodium nitrite, crystals for synthesis, Sigma-Aldrich®
Sodium nitrite
Formula:
NaNO₂ Boiling Pt: 320 °C (1013 hPa) Melting Pt: 284 °C Density: 2.168 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011118 CAS Number: 7632-00-0 EINECS: 231-555-9 UN: 1500 ADR: 5.1 (6.1),III Merck Index: 13,08721 |
Specification Test Results
Solubility | 820 g/l (20 °C) |
Melting Point | 280 °C (decomposition) |
Molar Mass | 69.00 g/mol |
Bulk Density | 1200 kg/m3 |
Boiling Point | 320 °C (decomposition) |
Density | 2.1 g/cm3 (20 °C) |
pH | 9 (100 g/l, H2O, 20 °C) |
Ignition Point | 510 °C |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...