Indole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1H-Indole

M-Clarity™ quality level = MQ200

8.22281.0025 8.22281.0100 8.22281.0025 8.22281.0100
EM8.22281.0025CS 4340.26 CAD
EM8.22281.0025 EM8.22281.0100 CA8.22281.0025 CA8.22281.0100
Indole for synthesis, Sigma-Aldrich®
Indole
Formula: C₈H₇N
Boiling Pt: 254…255 °C (1013 hPa)
Melting Pt: 50…51 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,04985

Order Now

Specification Test Results

Solubility 3.560 g/l (25 °C)
Melting Point 52 °C
Molar Mass 117.15 g/mol
Bulk Density 230 kg/m3
Boiling Point 254 °C
Vapor Pressure 0.016 hPa (25 °C)
Flash Point 110 °C
Density 1.22 g/cm3 (20 °C)
pH 5.9 (1000 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR