Acetic anhydride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Acetyl oxide, Acetyl acetate

M-Clarity™ quality level = MQ200

8.22278.1000 8.22278.2500 8.22278.9025
CA8.22278.1000EA 220 CAD
CA8.22278.1000 CA8.22278.2500 CA8.22278.9025
Acetic anhydride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Acetic anhydride
Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 139.9
Melting Pt: -73
Density: 1.08 g/cm3 (20 °C)
Flash Pt: 49
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8 II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) Hydrolysis,Risk of violent reaction.
Melting Point -73 °C
Molar Mass 102.08 g/mol
Boiling Point 138 - 140.5 °C (1013 hPa)
Vapor Pressure 4 hPa (20 °C)
Flash Point 49 °C
Explosion Limit 2.0 - 10.2 %(V)
Density 1.08 g/cm3 (20 °C)
pH 3 (10 g/l, H2O, 20 °C)
Ignition Point 330 °C
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR